missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Ethylenedioxythiophene-2,5-dicarboxylic Acid 95.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 230.19 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4-Ethylenedioxythiophene-2,5-dicarboxylic Acid |
Safety and Handling
| TSCA | No |