missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dinitrochlorobenzene 90.0+%, TCI America™

Catalog No. D190025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D190025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D190025G Supplier TCI America Supplier No. D190025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610-40-2
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00007212
InChI Key QVQSOXMXXFZAKU-UHFFFAOYSA-N
Synonym 1-chloro-3,4-dinitrobenzene, 3,4-dinitrochlorobenzene, benzene, 4-chloro-1,2-dinitro, unii-z29b1f274e, 1,2-dinitro-4-chlorobenzene, 1,2-dino2 4-cl benzene, 4-chlordinitrobenzol, acmc-209mog, 4-chloro-1,2-dinitrobenzen, ghl.pd_mitscher_leg0.928
PubChem CID 33097
IUPAC Name 4-chloro-1,2-dinitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 36°C
Boiling Point 315°C
Quantity 25 g
UN Number 3441
Formula Weight 202.55
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Dinitrochlorobenzene
EINECSNumber (3)-0454
TSCA Yes
Recommended Storage Refrigerator