missing translation for 'onlineSavingsMsg'
Learn More

3,4'-Dinitrobenzophenone 98.0+%, TCI America™

Catalog No. p-7127362
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25515G 5 g
D255125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25515G Supplier TCI America Supplier No. D25515G
Only null left

Chemical Identifiers

CAS 1469-74-5
Molecular Formula C13H8N2O5
Molecular Weight (g/mol) 272.216
MDL Number MFCD00617174
InChI Key ZEGCOKXUTZGBGN-UHFFFAOYSA-N
PubChem CID 12756333
IUPAC Name (3-nitrophenyl)-(4-nitrophenyl)methanone
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 173°C
Quantity 5 g
UN Number 1325
Formula Weight 272.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4′-Dinitrobenzophenone