missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dinitrobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America D082325G
Specifications
| 3,4-Dinitrobenzoic Acid | |
| 165°C | |
| C7H4N2O6 | |
| 25 g | |
| OMVRRHJJQILIJX-UHFFFAOYSA-N | |
| 3,4-dinitrobenzoic acid | |
| 10709 | |
| ≥98.0% (T) |
| 528-45-0 | |
| Yellow | |
| MFCD00007255 | |
| 3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956 | |
| OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O | |
| 212.12 | |
| 212.12 | |
| Crystalline Powder |
Chemical Identifiers
| 528-45-0 | |
| 212.12 | |
| OMVRRHJJQILIJX-UHFFFAOYSA-N | |
| 10709 | |
| OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O |
| C7H4N2O6 | |
| MFCD00007255 | |
| 3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956 | |
| 3,4-dinitrobenzoic acid |
Safety and Handling
EINECSNumber : (3)-1505
RTECSNumber : DG9140650
TSCA : Yes