missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dinitrobenzoic acid, 98+%

Catalog No. AAA1877414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1877414 25 g
AAA1877406 5 g
AAA1877422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1877414 Supplier Thermo Scientific Chemicals Supplier No. A1877414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 528-45-0 | C7H4N2O6 | 212.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-45-0
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007255
InChI Key OMVRRHJJQILIJX-UHFFFAOYSA-N
Synonym 3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956
PubChem CID 10709
IUPAC Name 3,4-dinitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 163°C to 167°C
Quantity 25 g
Beilstein 2055333
Merck Index 14,3275
Formula Weight 212.12
Percent Purity 99%
Chemical Name or Material 3,4-Dinitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-433-9
RTECSNumber DG9140650
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only