missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylthiophenol, 98%

Catalog No. AAB2316803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2316803 1 g
AAB2316806 5 g
AAB2316814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2316803 Supplier Thermo Scientific Chemicals Supplier No. B2316803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18800-53-8 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18800-53-8
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010023
InChI Key IDKCKPBAFOIONK-UHFFFAOYSA-N
Synonym 3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol
PubChem CID 87799
IUPAC Name 3,4-dimethylbenzenethiol
SMILES CC1=C(C=C(C=C1)S)C

Specifications

Density 1.027
Boiling Point 218°C
Flash Point 90°C (194°F)
Odor Unpleasant
Refractive Index 1.5736
Quantity 1 g
UN Number UN2810
Beilstein 2040353
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylthiophenol
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 242-587-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only