missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylthiobenzamide, 97%, Thermo Scientific Chemicals

Catalog No. p-7034135
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5179303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5179303 Supplier Thermo Scientific Chemicals Supplier No. H5179303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58952-03-7 | Molecular Formula: C9H11NS | Molecular Weight: 165.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58952-03-7
Molecular Formula C9H11NS
Molecular Weight (g/mol) 165.25
MDL Number MFCD06738326
InChI Key MGSZKKVFYBQMPF-UHFFFAOYSA-N
Synonym 3,4-dimethyl-thiobenzamide, 3,4-dimethylthiobenzamide, 3,4-dimethylbenzothioamide, 3,4-dimethylbenzene-1-carbothioamide, 3,4-dimethyl-thiobenzamid
PubChem CID 20470851
IUPAC Name 3,4-dimethylbenzenecarbothioamide
SMILES CC1=C(C)C=C(C=C1)C(N)=S

Specifications

Quantity 1 g
Formula Weight 165.25
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylthiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only