missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylnitrobenzene 98.0+%, TCI America™

Catalog No. D108525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D108525G 25 g
D1085500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D108525G Supplier TCI America Supplier No. D108525G

Chemical Identifiers

CAS 99-51-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007268
InChI Key HFZKOYWDLDYELC-UHFFFAOYSA-N
Synonym 4-nitro-o-xylene, p-nitro-o-xylene, o-xylene, 4-nitro, benzene, 1,2-dimethyl-4-nitro, para-nitro-ortho-xylene, 3,4-dimethyl-1-nitrobenzene, 4-nitro-1,2-dimethylbenzene, 1,2-dimethyl-4-nitrobenzol, 3,4-dimethylnitrobenzene, ccris 3118
PubChem CID 7440
IUPAC Name 1,2-dimethyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1C)[N+]([O-])=O

Specifications

Color Yellow
Quantity 25 g
Physical Form Crystal-Lump at 20°C
UN Number 3447
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3,4-Dimethylnitrobenzene
EINECSNumber (3)-0399
TSCA Yes