missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylhexane 97.0+%, TCI America™

Catalog No. D12285ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12285ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12285ML Supplier TCI America Supplier No. D12285ML

Chemical Identifiers

CAS 583-48-2
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00039927
InChI Key RNTWWGNZUXGTAX-UHFFFAOYSA-N
Synonym hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso
PubChem CID 11412
IUPAC Name 3,4-dimethylhexane
SMILES CCC(C)C(C)CC

Specifications

Color Colorless
Boiling Point 117°C
Quantity 5 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethylhexane
EINECSNumber (2)-0008
TSCA Yes