accessibility menu, dialog, popup

UserName

H3CCH3CH3CH3

CAS RN 583-48-2

IUPAC Name: 3,4-dimethylhexane
Synonyms: 3,4-dimethylhexane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: RNTWWGNZUXGTAX-UHFFFAOYNA-N
SMILES: CCC(C)C(C)CC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)