missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylbenzylamine 98.0+%, TCI America™
$511.68 - $511.68
Chemical Identifiers
| CAS | 102-48-7 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD00025577 |
| InChI Key | PXNRCZQMDSDSHJ-UHFFFAOYSA-N |
| PubChem CID | 66888 |
| IUPAC Name | (3,4-dimethylphenyl)methanamine |
| SMILES | CC1=C(C=C(C=C1)CN)C |
Chemical Identifiers
| 102-48-7 | |
| 135.21 | |
| PXNRCZQMDSDSHJ-UHFFFAOYSA-N | |
| (3,4-dimethylphenyl)methanamine |
| C9H13N | |
| MFCD00025577 | |
| 66888 | |
| CC1=C(C=C(C=C1)CN)C |
Specifications
| 102-48-7 | |
| 50°C | |
| MFCD00025577 | |
| 2735 | |
| PXNRCZQMDSDSHJ-UHFFFAOYSA-N | |
| (3,4-dimethylphenyl)methanamine | |
| 66888 | |
| ≥98.0% (GC,T) | |
| 3,4-Dimethylbenzylamine |
| Yellow | |
| C9H13N | |
| 25 mL | |
| 3,4-dimethylbenzylamine, 3,4-dimethylphenyl methanamine, benzenemethanamine, 3,4-dimethyl, benzylamine, 3,4-dimethyl, 1-3,4-dimethylphenyl methanamine, 3,4-dimethylbenzyl amine, 3,4-dimethyl-benzenemethanamin, 3,4-dimethylbenzylamine hcl, 3,4-dimethylphenyl methylamine, 3, 4-dimethylbenzylamine | |
| CC1=C(C=C(C=C1)CN)C | |
| 135.21 | |
| 135.21 | |
| Liquid |
Safety and Handling
TSCA : Yes