missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylbenzylamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 50°C |
| Quantity | 25 mL |
| UN Number | 2735 |
| Synonym | 3,4-dimethylbenzylamine, 3,4-dimethylphenyl methanamine, benzenemethanamine, 3,4-dimethyl, benzylamine, 3,4-dimethyl, 1-3,4-dimethylphenyl methanamine, 3,4-dimethylbenzyl amine, 3,4-dimethyl-benzenemethanamin, 3,4-dimethylbenzylamine hcl, 3,4-dimethylphenyl methylamine, 3, 4-dimethylbenzylamine |
| Formula Weight | 135.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3,4-Dimethylbenzylamine |
Safety and Handling
| TSCA | Yes |