missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylbenzyl Chloride (contains isomer) 70.0+%, TCI America™
Supplier: TCI America D029025G
Specifications
| 3,4-Dimethylbenzyl Chloride (contains isomer) | |
| Colorless | |
| C9H11Cl | |
| 25 g | |
| 3,4-dimethylbenzyl chloride, 4-chloromethyl-1,2-dimethylbenzene, benzene, 4-chloromethyl-1,2-dimethyl, benzene, 1-chloromethyl-3,4-dimethyl, 4-chloromethyl-o-xylene, 4-chloromethyl-1,2-dimethyl-benzene, 1-chloromethyl-3,4-dimethylbenzene, acmc-20anej, pubchem14890 | |
| CC1=C(C=C(C=C1)CCl)C | |
| 154.637 | |
| 154.64 | |
| Liquid |
| 102-46-5 | |
| 117°C | |
| MFCD00000910 | |
| 3265 | |
| UBQRAAXAHIKWRI-UHFFFAOYSA-N | |
| 4-(chloromethyl)-1,2-dimethylbenzene | |
| 66018 | |
| ≥70.0% (GC) |
Chemical Identifiers
| 102-46-5 | |
| 154.637 | |
| UBQRAAXAHIKWRI-UHFFFAOYSA-N | |
| 66018 | |
| CC1=C(C=C(C=C1)CCl)C |
| C9H11Cl | |
| MFCD00000910 | |
| 3,4-dimethylbenzyl chloride, 4-chloromethyl-1,2-dimethylbenzene, benzene, 4-chloromethyl-1,2-dimethyl, benzene, 1-chloromethyl-3,4-dimethyl, 4-chloromethyl-o-xylene, 4-chloromethyl-1,2-dimethyl-benzene, 1-chloromethyl-3,4-dimethylbenzene, acmc-20anej, pubchem14890 | |
| 4-(chloromethyl)-1,2-dimethylbenzene |
Safety and Handling
TSCA : Yes