missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzophenone, 99%

Catalog No. p-7025479
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AAB2096514 25 g
AAB2096522 100 g
2 options

Catalog No. AAB2096514

Supplier: Thermo Scientific Chemicals B2096514

Only null left

CAS: 2571-39-3 | C15H14O | 210.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2571-39-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00008525
InChI Key JENOLWCGNVWTJN-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzophenone, 3,4-dimethylphenyl phenyl methanone, methanone, 3,4-dimethylphenyl phenyl, 3,4-dimethylphenyl-phenylmethanone, 3,4-dimethylphenyl phenyl ketone, acmc-1cbaf, 3,4-dimethyl-benzophenone, ksc490c7t, benzophenone, 3,4-dimethyl
PubChem CID 75730
IUPAC Name (3,4-dimethylphenyl)-phenylmethanone
SMILES CC1=CC=C(C=C1C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 44°C to 47°C
Boiling Point 335°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1952043
Formula Weight 210.28
Percent Purity 99%
Chemical Name or Material 3,4-Dimethylbenzophenone
EINECSNumber 219-916-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only