missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethylbenzophenone-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. D29785G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29785G Supplier TCI America Supplier No. D29785G

Chemical Identifiers

CAS 2159-42-4
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
InChI Key AYVFSZDAFPVJOA-UHFFFAOYSA-N
Synonym 2-(3,4-Dimethylbenzoyl)benzoic Acid
PubChem CID 304868
IUPAC Name 2-(3,4-dimethylbenzoyl)benzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O)C

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 254.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dimethylbenzophenone-2-carboxylic Acid