missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzoic Acid 99.0+%, TCI America™

Catalog No. D143910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143910G 10 g
D143925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D143910G Supplier TCI America Supplier No. D143910G

Chemical Identifiers

CAS 619-04-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002524
InChI Key OPVAJFQBSDUNQA-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
PubChem CID 12073
ChEBI CHEBI:64818
IUPAC Name 3,4-dimethylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)C

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 10 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥99.0% (T)
Chemical Name or Material 3,4-Dimethylbenzoic Acid
RTECSNumber DG8734020
TSCA Yes