missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzoic acid, 98%

Catalog No. AAA1926014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1926014 25 g
AAA1926022 100 g
AAA1926036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1926014 Supplier Thermo Scientific Chemicals Supplier No. A1926014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 619-04-5 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-04-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002524
InChI Key OPVAJFQBSDUNQA-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
PubChem CID 12073
ChEBI CHEBI:64818
IUPAC Name 3,4-dimethylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)C

Specifications

Melting Point 164°C to 167°C
Quantity 25 g
Odor Odorless
Beilstein 907267
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylbenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-576-7
RTECSNumber DG8734020
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only