missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzenesulfonyl chloride, 98%

Catalog No. p-7027786
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2480606 5 g
AAB2480614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2480606 Supplier Thermo Scientific Chemicals Supplier No. B2480606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2905-30-8 | C8H9ClO2S | 204.668 g/mol

3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agent, bases, water/ moisture.

Chemical Identifiers

CAS 2905-30-8
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00625742
InChI Key DNMQPRPJIWTNAX-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzene-1-sulfonyl chloride, 3,4-dimethylbenzenesulfonylchloride, 3,4-dimethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethyl, 3,4-dimethylphenyl chlorosulfone, ksc562m4t, 3,4-dimethylphenylsulfonyl chloride, 3,4-dimethylbenzenesulphonyl chloride, 3,4-dimethyl benzenesulfonyl chloride
PubChem CID 3689743
IUPAC Name 3,4-dimethylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)C

Specifications

Quantity 5 g
Odor Acrid
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-337-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only