missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylanisole, 99%

Catalog No. AAB2273518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2273518 50 g
AAB2273530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2273518 Supplier Thermo Scientific Chemicals Supplier No. B2273518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4685-47-6 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 4685-47-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008396
InChI Key LVUBSVWMOWKPDJ-UHFFFAOYSA-N
Synonym 3,4-dimethylanisole, 4-methoxy-1,2-dimethyl-benzene, 4-methoxy-o-xylene, benzene, 4-methoxy-1,2-dimethyl, 3,4-dimethyl anisole, 1,2-dimethyl-4-methoxybenzene, 3,4-dimethylphenyl methyl ether, acmc-20anel, pubchem5361
PubChem CID 78411
IUPAC Name 4-methoxy-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)OC)C

Specifications

Density 0.975
Quantity 50 g
Refractive Index 1.5175
Beilstein 2040907
Boiling Point 200°C to 201°C
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3,4-Dimethylanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 225-142-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only