missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylaniline 99.0+%, TCI America™

Catalog No. D0670500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0670500G 500 g
D067025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0670500G Supplier TCI America Supplier No. D0670500G

Chemical Identifiers

CAS 95-64-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007810
InChI Key DOLQYFPDPKPQSS-UHFFFAOYSA-N
Synonym 3,4-xylidine, 4-amino-o-xylene, 3,4-xylylamine, 3,4-dimethylphenylamine, benzenamine, 3,4-dimethyl, 3,4-dimethylbenzenamine, 3,4-dimethylaminobenzene, 1-amino-3,4-dimethylbenzene, 4-amino-1,2-dimethylbenzene, aniline, 3,4-dimethyl
PubChem CID 7248
ChEBI CHEBI:39901
IUPAC Name 3,4-dimethylaniline
SMILES CC1=CC=C(N)C=C1C

Specifications

Melting Point 51°C
Quantity 500 g
Physical Form Crystal-Powder at 20°C
UN Number 3452
Boiling Point 226°C
Formula Weight 121.18
Percent Purity ≥99.0% (GC,T)
Chemical Name or Material 3,4-Dimethylaniline
EINECSNumber (3)-0129
RTECSNumber ZE9450000
TSCA Yes