missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-o-phenylenediamine, 98%

CAS: 41927-01-9 | C8H12N2 | 136.198 g/mol

$108.04 - $379.53

Chemical Identifiers

CAS 41927-01-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00051709
InChI Key MHQULXYNBKWNDF-UHFFFAOYSA-N
Synonym 3,4-dimethyl-o-phenylenediamine, 2-amino-3,4-dimethylphenylamine, 2-amino-3,4-dimethylphenyl amine, 3,4-dimethyl-benzene-1,2-diamine, 1,2-benzenediamine, 3,4-dimethyl, 3,4-diamino-o-xylene, o-xylene-3,4-diamine, 5,6-dimethyl phenylenediamine, 1,2-diamino-3,4-dimethylbenzene, 3,4-dimethyl-1,2-benzenediamine
PubChem CID 521077
IUPAC Name 3,4-dimethylbenzene-1,2-diamine
SMILES CC1=C(C(=C(C=C1)N)N)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0745503
View Documents
Thermo Scientific Chemicals
L0745503
1 g
Each for $108.04
Only null left
 
AAL0745506
View Documents
Thermo Scientific Chemicals
L0745506
5 g
Each for $379.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41927-01-9
136.198
MHQULXYNBKWNDF-UHFFFAOYSA-N
521077
CC1=C(C(=C(C=C1)N)N)C
C8H12N2
MFCD00051709
3,4-dimethyl-o-phenylenediamine, 2-amino-3,4-dimethylphenylamine, 2-amino-3,4-dimethylphenyl amine, 3,4-dimethyl-benzene-1,2-diamine, 1,2-benzenediamine, 3,4-dimethyl, 3,4-diamino-o-xylene, o-xylene-3,4-diamine, 5,6-dimethyl phenylenediamine, 1,2-diamino-3,4-dimethylbenzene, 3,4-dimethyl-1,2-benzenediamine
3,4-dimethylbenzene-1,2-diamine

Specifications

86°C to 90°C
1 g
MFCD00051709
2716459
MHQULXYNBKWNDF-UHFFFAOYSA-N
3,4-dimethylbenzene-1,2-diamine
521077
98%
41927-01-9
C8H12N2
UN2811
3,4-dimethyl-o-phenylenediamine, 2-amino-3,4-dimethylphenylamine, 2-amino-3,4-dimethylphenyl amine, 3,4-dimethyl-benzene-1,2-diamine, 1,2-benzenediamine, 3,4-dimethyl, 3,4-diamino-o-xylene, o-xylene-3,4-diamine, 5,6-dimethyl phenylenediamine, 1,2-diamino-3,4-dimethylbenzene, 3,4-dimethyl-1,2-benzenediamine
CC1=C(C(=C(C=C1)N)N)C
136.198
136.2
3,4-Dimethyl-o-phenylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only