missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-o-phenylenediamine, 98%

Catalog No. AAL0745506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0745506 5 g
AAL0745503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0745506 Supplier Thermo Scientific Chemicals Supplier No. L0745506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 41927-01-9 | C8H12N2 | 136.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41927-01-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00051709
InChI Key MHQULXYNBKWNDF-UHFFFAOYSA-N
Synonym 3,4-dimethyl-o-phenylenediamine, 2-amino-3,4-dimethylphenylamine, 2-amino-3,4-dimethylphenyl amine, 3,4-dimethyl-benzene-1,2-diamine, 1,2-benzenediamine, 3,4-dimethyl, 3,4-diamino-o-xylene, o-xylene-3,4-diamine, 5,6-dimethyl phenylenediamine, 1,2-diamino-3,4-dimethylbenzene, 3,4-dimethyl-1,2-benzenediamine
PubChem CID 521077
IUPAC Name 3,4-dimethylbenzene-1,2-diamine
SMILES CC1=C(C(=C(C=C1)N)N)C

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
UN Number UN2811
Beilstein 2716459
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 3,4-Dimethyl-o-phenylenediamine
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only