missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethyl-1,2-cyclopentanedione 98.0+%, TCI America™
$119.30 - $119.30
Chemical Identifiers
| CAS | 13494-06-9 |
|---|---|
| Molecular Formula | C7H10O2 |
| Molecular Weight (g/mol) | 126.155 |
| MDL Number | MFCD00143063 |
| InChI Key | WGAVDEVFJDQIMZ-UHFFFAOYSA-N |
| Synonym | 3,4-dimethyl-1,2-cyclopentanedione, 3,4-dimethyl-1,2-cyclopentadione, 1,2-cyclopentanedione, 3,4-dimethyl, fema no. 3268, 1,2-cyclopentanedione,3,4-dimethyl, caramel pentadione, 3,4-dimethyl 1,2-cyclopentandione, acmc-209bxe, 3,4-dimethyl-1,2-cyclopentadione, fg |
| PubChem CID | 61633 |
| IUPAC Name | 3,4-dimethylcyclopentane-1,2-dione |
| SMILES | CC1CC(=O)C(=O)C1C |
Chemical Identifiers
| 13494-06-9 | |
| 126.155 | |
| WGAVDEVFJDQIMZ-UHFFFAOYSA-N | |
| 61633 | |
| CC1CC(=O)C(=O)C1C |
| C7H10O2 | |
| MFCD00143063 | |
| 3,4-dimethyl-1,2-cyclopentanedione, 3,4-dimethyl-1,2-cyclopentadione, 1,2-cyclopentanedione, 3,4-dimethyl, fema no. 3268, 1,2-cyclopentanedione,3,4-dimethyl, caramel pentadione, 3,4-dimethyl 1,2-cyclopentandione, acmc-209bxe, 3,4-dimethyl-1,2-cyclopentadione, fg | |
| 3,4-dimethylcyclopentane-1,2-dione |
Specifications
| 13494-06-9 | |
| C7H10O2 | |
| 1 g | |
| WGAVDEVFJDQIMZ-UHFFFAOYSA-N | |
| 3,4-dimethylcyclopentane-1,2-dione | |
| 61633 | |
| ≥98.0% (T) | |
| 3,4-Dimethyl-1,2-cyclopentanedione |
| 69°C | |
| MFCD00143063 | |
| 3,4-dimethyl-1,2-cyclopentanedione, 3,4-dimethyl-1,2-cyclopentadione, 1,2-cyclopentanedione, 3,4-dimethyl, fema no. 3268, 1,2-cyclopentanedione,3,4-dimethyl, caramel pentadione, 3,4-dimethyl 1,2-cyclopentandione, acmc-209bxe, 3,4-dimethyl-1,2-cyclopentadione, fg | |
| CC1CC(=O)C(=O)C1C | |
| 126.155 | |
| 126.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes