accessibility menu, dialog, popup

UserName

CAS RN 13494-06-9

CH3OOH3C

CAS RN 13494-06-9

IUPAC Name: 3,4-dimethylcyclopentane-1,2-dione
Synonyms: 3,4-dimethylcyclopentane-1,2-dione
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: WGAVDEVFJDQIMZ-UHFFFAOYNA-N
SMILES: CC1CC(=O)C(=O)C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results