missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxythiophene, 98%

CAS: 51792-34-8 | C6H8O2S | 144.188 g/mol

Supplier:  Thermo Scientific Chemicals H5667406

 View more versions of this product

Catalog No. AAH5667406


Only null left
Explore available promotions

Description

Description

3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

3,4-Dimethoxythiophene
1.209
106°C (223°F)
1.5409
5 g
Miscible with organic solvents. Slightly miscible with water.
COC1=CSC=C1OC
144.188
144.19
51792-34-8
100°C to 102°C (10 mmHg to 11 mmHg)
C6H8O2S
MFCD01096546
3,4-dimethoxy thiophene, thiophene, 3,4-dimethoxy, pubchem15752, 3,4-dimethoxy-thiophene, acmc-209kw1, ksc491i4n, 3,4-dimethoxythiophene
ZUDCKLVMBAXBIF-UHFFFAOYSA-N
3,4-dimethoxythiophene
3613501
98%

Chemical Identifiers

51792-34-8
144.188
ZUDCKLVMBAXBIF-UHFFFAOYSA-N
3613501
COC1=CSC=C1OC
C6H8O2S
MFCD01096546
3,4-dimethoxy thiophene, thiophene, 3,4-dimethoxy, pubchem15752, 3,4-dimethoxy-thiophene, acmc-209kw1, ksc491i4n, 3,4-dimethoxythiophene
3,4-dimethoxythiophene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only