missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxythiophene, 98%

Catalog No. p-7037848
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5667403 1 g
AAH5667406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5667403 Supplier Thermo Scientific Chemicals Supplier No. H5667403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51792-34-8 | C6H8O2S | 144.188 g/mol

3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 51792-34-8
Molecular Formula C6H8O2S
Molecular Weight (g/mol) 144.188
MDL Number MFCD01096546
InChI Key ZUDCKLVMBAXBIF-UHFFFAOYSA-N
Synonym 3,4-dimethoxy thiophene, thiophene, 3,4-dimethoxy, pubchem15752, 3,4-dimethoxy-thiophene, acmc-209kw1, ksc491i4n, 3,4-dimethoxythiophene
PubChem CID 3613501
IUPAC Name 3,4-dimethoxythiophene
SMILES COC1=CSC=C1OC

Specifications

Density 1.209
Boiling Point 100°C to 102°C (10 mmHg to 11 mmHg)
Flash Point 106°C (223°F)
Refractive Index 1.5409
Quantity 1 g
Solubility Information Miscible with organic solvents. Slightly miscible with water.
Formula Weight 144.19
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxythiophene
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only