missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetonitrile, 98%

CAS: 93-17-4 | C10H11NO2 | 177.203 g/mol

Supplier:  Thermo Scientific Chemicals A1200322

 View more versions of this product

Catalog No. AAA1200322


Only null left
Explore available promotions

Description

Description

It is employed as an intermediate in the preparation of the muscle relaxant papverin. 3,4-Dimethoxyphenyl)acetonitrile has been used in the synthesis of (Z)-3-(benzo[b]thiophen-2-yl)-2-(3,4-dimethoxyphenyl)acrylonitrile1. It has also been used in the preparation of modified diterpene (±) nimbidiol which relies on a sulfenium ion promoted polyene cyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the preparation of the muscle relaxant papverin. 3,4-Dimethoxyphenyl)acetonitrile has been used in the synthesis of (Z)-3-(benzo[b]thiophen-2-yl)-2-(3,4-dimethoxyphenyl)acrylonitrile1. It has also been used in the preparation of modified diterpene (±) nimbidiol which relies on a sulfenium ion promoted polyene cyclization.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

3,4-Dimethoxyphenylacetonitrile
62°C to 65°C
Characteristic
MFCD00001911
1956100
Insoluble in water. Soluble in methanol.
COC1=C(C=C(C=C1)CC#N)OC
177.203
177.2
93-17-4
177°C to 178°C (10 mmHg)
C10H11NO2
100 g
3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
ASLSUMISAQDOOB-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)acetonitrile
66727
98%

Chemical Identifiers

93-17-4
177.203
ASLSUMISAQDOOB-UHFFFAOYSA-N
66727
COC1=C(C=C(C=C1)CC#N)OC
C10H11NO2
MFCD00001911
3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
2-(3,4-dimethoxyphenyl)acetonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 202-225-1

RTECSNumber : AL9325000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only