missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzyl Cyanide 98.0+%, TCI America™

Catalog No. p-7126992
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D188725G 25 g
D1887500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D188725G Supplier TCI America Supplier No. D188725G

Chemical Identifiers

CAS 93-17-4
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00001911
InChI Key ASLSUMISAQDOOB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
PubChem CID 66727
IUPAC Name 2-(3,4-dimethoxyphenyl)acetonitrile
SMILES COC1=C(C=C(C=C1)CC#N)OC

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 177.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxybenzyl Cyanide
EINECSNumber (3)-1795
RTECSNumber AL9325000
TSCA No