missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxybenzoyl Chloride 98.0+%, TCI America™
$242.33 - $725.11
Chemical Identifiers
| CAS | 3535-37-3 |
|---|---|
| Molecular Formula | C9H9ClO3 |
| Molecular Weight (g/mol) | 200.618 |
| MDL Number | MFCD00000674 |
| InChI Key | VIOBGCWEHLRBEP-UHFFFAOYSA-N |
| Synonym | veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t |
| PubChem CID | 77070 |
| IUPAC Name | 3,4-dimethoxybenzoyl chloride |
| SMILES | COC1=C(C=C(C=C1)C(=O)Cl)OC |
Chemical Identifiers
| 3535-37-3 | |
| 200.618 | |
| VIOBGCWEHLRBEP-UHFFFAOYSA-N | |
| 77070 | |
| COC1=C(C=C(C=C1)C(=O)Cl)OC |
| C9H9ClO3 | |
| MFCD00000674 | |
| veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t | |
| 3,4-dimethoxybenzoyl chloride |
Specifications
| 3535-37-3 | |
| White | |
| MFCD00000674 | |
| 3261 | |
| VIOBGCWEHLRBEP-UHFFFAOYSA-N | |
| 3,4-dimethoxybenzoyl chloride | |
| 77070 | |
| ≥98.0% (GC,T) | |
| 3,4-Dimethoxybenzoyl Chloride |
| 72°C | |
| C9H9ClO3 | |
| 5 g | |
| veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t | |
| COC1=C(C=C(C=C1)C(=O)Cl)OC | |
| 200.618 | |
| 200.62 | |
| Crystalline Powder |
Safety and Handling
TSCA : No