missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzonitrile 98.0+%, TCI America™

Catalog No. D287025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D287025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D287025G Supplier TCI America Supplier No. D287025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2024-83-1
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00001802
InChI Key OSEQIDSFSBWXRE-UHFFFAOYSA-N
Synonym veratronitrile, 4-cyanoveratrole, 3,4-dimethoxy-benzonitrile, 4-cyano-1,2-dimethoxybenzene, 1-cyano-3,4-dimethoxylbenzene, 3,4-dimethoxybenzoic acid nitrile, 3,4-dimethoxyphenyl cyanide, benzonitrile,4-dimethoxy, 3,4-dimethoxybenzenecarbonitrile, 3-cyanoveratrole
PubChem CID 74842
IUPAC Name 3,4-dimethoxybenzonitrile
SMILES COC1=CC=C(C=C1OC)C#N

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxybenzonitrile