missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™
$61.61 - $683.44
Chemical Identifiers
| CAS | 5222-73-1 |
|---|---|
| Molecular Formula | C6H6O4 |
| Molecular Weight (g/mol) | 142.11 |
| MDL Number | MFCD00101316 |
| InChI Key | SZBNZTGCAMLMJY-UHFFFAOYSA-N |
| PubChem CID | 123227 |
| IUPAC Name | 3,4-dimethoxycyclobut-3-ene-1,2-dione |
| SMILES | COC1=C(C(=O)C1=O)OC |
Chemical Identifiers
| 5222-73-1 | |
| 142.11 | |
| SZBNZTGCAMLMJY-UHFFFAOYSA-N | |
| 3,4-dimethoxycyclobut-3-ene-1,2-dione |
| C6H6O4 | |
| MFCD00101316 | |
| 123227 | |
| COC1=C(C(=O)C1=O)OC |
Specifications
| 5222-73-1 | |
| White | |
| MFCD00101316 | |
| 3,4-dimethoxy-3-cyclobutene-1,2-dione, dimethyl squarate, squaric acid dimethyl ester, 3-cyclobutene-1,2-dione, 3,4-dimethoxy, 3,4-dimethoxy-cyclobut-3-ene-1,2-dione, dimethoxycyclobut-3-ene-1,2-dione, 3-cyclobutene-1,2-dione,3,4-dimethoxy, 3,4-dimethoxycyclobut-3-en-1,2-dion, dimethoxycyclobutenedione, pubchem13709 | |
| COC1=C(C(=O)C1=O)OC | |
| 142.11 | |
| 142.11 | |
| Crystalline Powder |
| 56°C | |
| C6H6O4 | |
| 1 g | |
| SZBNZTGCAMLMJY-UHFFFAOYSA-N | |
| 3,4-dimethoxycyclobut-3-ene-1,2-dione | |
| 123227 | |
| ≥98.0% (GC) | |
| 3,4-Dimethoxy-3-cyclobutene-1,2-dione |
Safety and Handling
TSCA : No