accessibility menu, dialog, popup

UserName

H3COOCH3OO

CAS RN 5222-73-1

IUPAC Name: dimethoxycyclobut-3-ene-1,2-dione
Synonyms: dimethoxycyclobut-3-ene-1,2-dione
Molecular Weight (g/mol): 142.11
Molecular Formula: C6H6O4
InChi Key: SZBNZTGCAMLMJY-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(=O)C1=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)