missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™

Catalog No. D20991G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20991G 1 g
D20995G 5 g
D209925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D20991G Supplier TCI America Supplier No. D20991G

Chemical Identifiers

CAS 5222-73-1
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00101316
InChI Key SZBNZTGCAMLMJY-UHFFFAOYSA-N
PubChem CID 123227
IUPAC Name 3,4-dimethoxycyclobut-3-ene-1,2-dione
SMILES COC1=C(C(=O)C1=O)OC

Specifications

Melting Point 56°C
Color White
Quantity 1 g
Synonym 3,4-dimethoxy-3-cyclobutene-1,2-dione, dimethyl squarate, squaric acid dimethyl ester, 3-cyclobutene-1,2-dione, 3,4-dimethoxy, 3,4-dimethoxy-cyclobut-3-ene-1,2-dione, dimethoxycyclobut-3-ene-1,2-dione, 3-cyclobutene-1,2-dione,3,4-dimethoxy, 3,4-dimethoxycyclobut-3-en-1,2-dion, dimethoxycyclobutenedione, pubchem13709
Formula Weight 142.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxy-3-cyclobutene-1,2-dione