Learn More
3,4-Dimethoxy-3-cyclobutene-1,2-dione, 98%
CAS: 5222-73-1 | C6H6O4 | 142.11 g/mol
$103.11 - $333.64
Chemical Identifiers
| CAS | 5222-73-1 |
|---|---|
| Molecular Formula | C6H6O4 |
| Molecular Weight (g/mol) | 142.11 |
| MDL Number | MFCD00101316 |
| InChI Key | SZBNZTGCAMLMJY-UHFFFAOYSA-N |
| PubChem CID | 123227 |
| IUPAC Name | 3,4-dimethoxycyclobut-3-ene-1,2-dione |
| SMILES | COC1=C(C(=O)C1=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1818603
|
Thermo Scientific Chemicals
A1818603 |
1 g |
Each for $103.11
|
|
|||||
|
AAA1818606
|
Thermo Scientific Chemicals
A1818606 |
5 g |
Each for $333.64
|
|
|||||
Description
3,4-Dimethoxy-3-cyclobutene-1,2-dione (Dimethyl squarate) finds it uses in the synthesis of chiral squaramides, highly enantioselective catalyst for the Friedel-Crafts reactions of indoles, 3-(hydroxyamino)-4-methoxy-3-cyclobutene-1,2-dione, squarate derivatives of the O-SP-core antigens, methyl squarate derivative of the Ogawa O-SP-core antigen, o-quinodimethanes, benzocyclobutenes, quinones. It is employed as a starting material in the preparation of o-quinodimethanes, benzocyclobutenes, and quinones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethoxy-3-cyclobutene-1,2-dione (Dimethyl squarate) finds it uses in the synthesis of chiral squaramides, highly enantioselective catalyst for the Friedel-Crafts reactions of indoles, 3-(hydroxyamino)-4-methoxy-3-cyclobutene-1,2-dione, squarate derivatives of the O-SP-core antigens, methyl squarate derivative of the Ogawa O-SP-core antigen, o-quinodimethanes, benzocyclobutenes, quinones. It is employed as a starting material in the preparation of o-quinodimethanes, benzocyclobutenes, and quinones.
Solubility
Very soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 5222-73-1 | |
| 142.11 | |
| SZBNZTGCAMLMJY-UHFFFAOYSA-N | |
| 3,4-dimethoxycyclobut-3-ene-1,2-dione |
| C6H6O4 | |
| MFCD00101316 | |
| 123227 | |
| COC1=C(C(=O)C1=O)OC |
Specifications
| 5222-73-1 | |
| C6H6O4 | |
| 1 g | |
| Very soluble in water. | |
| COC1=C(C(=O)C1=O)OC | |
| 142.11 | |
| 142.1 | |
| 3,4-Dimethoxy-3-cyclobutene-1,2-dione |
| 53°C to 56°C | |
| MFCD00101316 | |
| 3,4-dimethoxy-3-cyclobutene-1,2-dione, dimethyl squarate, squaric acid dimethyl ester, 3-cyclobutene-1,2-dione, 3,4-dimethoxy, 3,4-dimethoxy-cyclobut-3-ene-1,2-dione, dimethoxycyclobut-3-ene-1,2-dione, 3-cyclobutene-1,2-dione,3,4-dimethoxy, 3,4-dimethoxycyclobut-3-en-1,2-dion, dimethoxycyclobutenedione, pubchem13709 | |
| SZBNZTGCAMLMJY-UHFFFAOYSA-N | |
| 3,4-dimethoxycyclobut-3-ene-1,2-dione | |
| 123227 | |
| 98% |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only