missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydroxyphenylacetic acid, ≥97.0% (HPLC); 97.0-103.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 102-32-9 |
| Physical Form | Neat |
| Linear Formula | (HO)2C6H3 CH2 CO2 H |
| MDL Number | MFCD00004338 |
| Melting Point | 127°C to 130°C |
| Synonym | DOPAC; Homoprotocatechuic acid |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 168.15 |
| Percent Purity | ≥97.0% (HPLC); 97.0 to 103.0% (T) |
| Show More |