missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4'-Dihydroxyflavone 97.0+%, TCI America™
$229.10 - $229.10
Chemical Identifiers
| CAS | 14919-49-4 |
|---|---|
| Molecular Formula | C15H10O4 |
| Molecular Weight (g/mol) | 254.241 |
| MDL Number | MFCD00017690 |
| InChI Key | GPGOCTLAUAHUQO-UHFFFAOYSA-N |
| Synonym | 3,4'-dihydroxyflavone, 3-hydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, 3-hydroxy-2-4-hydroxyphenyl chromen-4-one, 3-hydroxy-2-4-hydroxy-phenyl-chromen-4-one, 4'-hydroxyflavonol, acmc-20anf1, 3,4/'-dihydroxyflavone, spectrum1505138, 3-hydroxy-2-4-hydroxyphenyl chromone, 4h-1-benzopyran-4-one,3-hydroxy-2-4-hydroxyphenyl |
| PubChem CID | 688715 |
| IUPAC Name | 3-hydroxy-2-(4-hydroxyphenyl)chromen-4-one |
| SMILES | C1=CC=C2C(=C1)C(=O)C(=C(O2)C3=CC=C(C=C3)O)O |
Chemical Identifiers
| 14919-49-4 | |
| 254.241 | |
| GPGOCTLAUAHUQO-UHFFFAOYSA-N | |
| 688715 | |
| C1=CC=C2C(=C1)C(=O)C(=C(O2)C3=CC=C(C=C3)O)O |
| C15H10O4 | |
| MFCD00017690 | |
| 3,4'-dihydroxyflavone, 3-hydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, 3-hydroxy-2-4-hydroxyphenyl chromen-4-one, 3-hydroxy-2-4-hydroxy-phenyl-chromen-4-one, 4'-hydroxyflavonol, acmc-20anf1, 3,4/'-dihydroxyflavone, spectrum1505138, 3-hydroxy-2-4-hydroxyphenyl chromone, 4h-1-benzopyran-4-one,3-hydroxy-2-4-hydroxyphenyl | |
| 3-hydroxy-2-(4-hydroxyphenyl)chromen-4-one |
Specifications
| 14919-49-4 | |
| C15H10O4 | |
| 1 g | |
| GPGOCTLAUAHUQO-UHFFFAOYSA-N | |
| 3-hydroxy-2-(4-hydroxyphenyl)chromen-4-one | |
| 688715 | |
| ≥97.0% (GC) | |
| 3,4′-Dihydroxyflavone |
| 275°C | |
| MFCD00017690 | |
| 3,4'-dihydroxyflavone, 3-hydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, 3-hydroxy-2-4-hydroxyphenyl chromen-4-one, 3-hydroxy-2-4-hydroxy-phenyl-chromen-4-one, 4'-hydroxyflavonol, acmc-20anf1, 3,4/'-dihydroxyflavone, spectrum1505138, 3-hydroxy-2-4-hydroxyphenyl chromone, 4h-1-benzopyran-4-one,3-hydroxy-2-4-hydroxyphenyl | |
| C1=CC=C2C(=C1)C(=O)C(=C(O2)C3=CC=C(C=C3)O)O | |
| 254.241 | |
| 254.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No