missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4'-Dihydroxyflavone 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 275°C |
| Quantity | 1 g |
| Formula Weight | 254.24 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4′-Dihydroxyflavone |
Safety and Handling
| TSCA | No |