Learn More
3,4-Dihydroxycinnamic acid, predominantly trans, 98+%
CAS: 331-39-5 | C9H8O4 | 180.159 g/mol
Supplier: Thermo Scientific Chemicals A1595009
Description
3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.
Solubility
Soluble in DMSO (40 mg/mol), ethanol (25 mg/mL warm), DMF (∼7 mg/mL), PBS, PH 7.2 (∼0.65 mg/mol), hot water, ethyl acetate, Chloroform Hexane, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications
| 3,4-Dihydroxycinnamic acid | |
| 331-39-5 | |
| C9H8O4 | |
| 10 g | |
| 14,1635 | |
| Soluble in DMSO (40mg/mol),ethanol (25mg/mL warm),DMF (∽7mg/mL),PBS,PH 7.2 (∽0.65mg/mol),hot water,ethyl acetate,Chloroform Hexane,and methanol. | |
| C1=CC(=C(C=C1C=CC(=O)O)O)O | |
| 180.159 | |
| CHEBI:16433 | |
| 99% |
| predominantly trans | |
| ∼202°C (decomposition) | |
| MFCD00004392 | |
| 2210883 | |
| caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid | |
| QAIPRVGONGVQAS-DUXPYHPUSA-N | |
| (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid | |
| 689043 | |
| 180.16 |
Chemical Identifiers
| 331-39-5 | |
| 180.159 | |
| QAIPRVGONGVQAS-DUXPYHPUSA-N | |
| 689043 | |
| (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid |
| C9H8O4 | |
| MFCD00004392 | |
| caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid | |
| CHEBI:16433 | |
| C1=CC(=C(C=C1C=CC(=O)O)O)O |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H351-H361
EINECSNumber : 206-361-2
RTECSNumber : GD8950000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only