missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzyl Alcohol 96.0+%, TCI America™

Catalog No. p-7127406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26201G 1 g
D26205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26201G Supplier TCI America Supplier No. D26201G
Only null left

Chemical Identifiers

CAS 3897-89-0
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00210409
InChI Key PCYGLFXKCBFGPC-UHFFFAOYSA-N
Synonym 4-Hydroxymethylpyrocatechol, alpha,3,4-Trihydroxytoluene
PubChem CID 100733
ChEBI CHEBI:20412
IUPAC Name 4-(hydroxymethyl)benzene-1,2-diol
SMILES OCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxybenzyl Alcohol
TSCA No
Recommended Storage Freezer