missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydroxybenzyl Alcohol 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 117°C |
| Quantity | 1 g |
| Formula Weight | 140.14 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4-Dihydroxybenzyl Alcohol |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |