missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzophenone 98.0+%, TCI America™

Catalog No. D33475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33475G 5 g
D334725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33475G Supplier TCI America Supplier No. D33475G
Only null left

Chemical Identifiers

CAS 10425-11-3
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00477203
InChI Key ARWCZKJISXFBGI-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzophenone, 3,4-dihydroxyphenyl phenyl methanone, 3,4-dihydroxyphenyl phenylmethanone, methanone, 3,4-dihydroxyphenyl phenyl, 3,4-dihydroxy-diphenyl ketone, 3,4-dihydroxyphenyl-phenylmethanone, 4-benzoylbenzene-1,2-diol, 3,4-dihydroxyphenyl phenyl ketone, 4-benzoylpyrocatechol, pubchem3385
PubChem CID 165870
IUPAC Name (3,4-dihydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)O)O

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxybenzophenone