missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzhydrazide, 97%

Catalog No. p-7025986
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2174606 5 g
AAB2174614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2174606 Supplier Thermo Scientific Chemicals Supplier No. B2174606

CAS: 39635-11-5 | C7H8N2O3 | 168.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39635-11-5
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00017064
InChI Key WGXWEXNJRZMIPT-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzhydrazide, 3,4-dihydroxy-benzoic acid hydrazide, 4-???benzene-1,2-diol, protocatechuhydrazide, acmc-1ad2t, 3,4-dihydroxy-benzohydrazide, 3,4-bis oxidanyl benzohydrazide, 3,4-dihydroxybenzoic acid hydrazide, 3,4-dihydroxy-benzoesaeure-hydrazid, 3,4-dihydroxybenzoic acid, hydrazide
PubChem CID 586300
IUPAC Name 3,4-dihydroxybenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)O)O

Specifications

Melting Point ∼265°C (decomposition)
Quantity 5 g
Beilstein 2642968
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,4-Dihydroxybenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-550-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only