missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',4'-Dihydroxyacetophenone 98.0+%, TCI America™
Supplier: TCI America D234525G
Specifications
| 3′,4′-Dihydroxyacetophenone | |
| 119°C | |
| C8H8O3 | |
| 25 g | |
| UCQUAMAQHHEXGD-UHFFFAOYSA-N | |
| 1-(3,4-dihydroxyphenyl)ethanone | |
| 14530 | |
| 152.15 | |
| Crystalline Powder |
| 1197-09-7 | |
| Yellow | |
| MFCD00016439 | |
| 4-Acetocatechol, 4-Acetylpyrocatechol | |
| CC(=O)C1=CC(=C(C=C1)O)O | |
| 152.149 | |
| CHEBI:19868 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1197-09-7 | |
| 152.149 | |
| UCQUAMAQHHEXGD-UHFFFAOYSA-N | |
| 14530 | |
| 1-(3,4-dihydroxyphenyl)ethanone |
| C8H8O3 | |
| MFCD00016439 | |
| 4-Acetocatechol, 4-Acetylpyrocatechol | |
| CHEBI:19868 | |
| CC(=O)C1=CC(=C(C=C1)O)O |
Safety and Handling
EINECSNumber : (3)-1250&(3)-2941
RTECSNumber : KM5775200
TSCA : No