missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxyacetophenone 98.0+%, TCI America™

Catalog No. p-7127246
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23455G 5 g
D234525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23455G Supplier TCI America Supplier No. D23455G

Chemical Identifiers

CAS 1197-09-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00016439
InChI Key UCQUAMAQHHEXGD-UHFFFAOYSA-N
Synonym 4-Acetocatechol, 4-Acetylpyrocatechol
PubChem CID 14530
ChEBI CHEBI:19868
IUPAC Name 1-(3,4-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)O

Specifications

Melting Point 119°C
Color Yellow
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dihydroxyacetophenone
EINECSNumber (3)-1250&(3)-2941
RTECSNumber KM5775200
TSCA No