missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran-2-methanol 97.0+%, TCI America™

Catalog No. p-7127653
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32951G 1 g
D32955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32951G Supplier TCI America Supplier No. D32951G

Chemical Identifiers

CAS 3749-36-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
InChI Key XMICBFRKICBBKD-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-pyran-2-methanol, 2-hydroxymethyl-3,4-dihydro-2h-pyran, dhp linker, 3,4-dihydro-2h-pyran-2-yl methanol, 2h-pyran-2-methanol, 3,4-dihydro, dhp resin, ellmans dhp resin, zlchem 376, pubchem9703, acmc-209itf
PubChem CID 95559
IUPAC Name 3,4-dihydro-2H-pyran-2-ylmethanol
SMILES C1CC(OC=C1)CO

Specifications

Color Colorless
Boiling Point 93°C
Quantity 1 g
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dihydro-2H-pyran-2-methanol