accessibility menu, dialog, popup

UserName

HOO

CAS RN 3749-36-8

IUPAC Name: (3,4-dihydro-2H-pyran-2-yl)methanol
Synonyms: 3,4-dihydro-2H-pyran-2-ylmethanol
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: XMICBFRKICBBKD-UHFFFAOYNA-N
SMILES: OCC1CCC=CO1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)