missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran-2-methanol, 96%

Catalog No. p-7037401
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5612403 1 g
AAH5612406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5612403 Supplier Thermo Scientific Chemicals Supplier No. H5612403

CAS: 3749-36-8 | C6H10O2 | 114.144 g/mol

It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

Notes
Incompatible with oxidizing agents, acids and heats. Store at room temperature.

Chemical Identifiers

CAS 3749-36-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00051507
InChI Key XMICBFRKICBBKD-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-pyran-2-methanol, 2-hydroxymethyl-3,4-dihydro-2h-pyran, dhp linker, 3,4-dihydro-2h-pyran-2-yl methanol, 2h-pyran-2-methanol, 3,4-dihydro, dhp resin, ellmans dhp resin, zlchem 376, pubchem9703, acmc-209itf
PubChem CID 95559
IUPAC Name 3,4-dihydro-2H-pyran-2-ylmethanol
SMILES C1CC(OC=C1)CO

Specifications

Density 1.101
Boiling Point 92°C to 93°C (25 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.478
Quantity 1 g
Formula Weight 114.14
Percent Purity 96%
Chemical Name or Material 3,4-Dihydro-2H-pyran-2-methanol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 223-150-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only