missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxylic acid, 98%

Catalog No. AAH2717506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2717506 5 g
AAH2717503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2717506 Supplier Thermo Scientific Chemicals Supplier No. H2717506

CAS: 33632-74-5 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33632-74-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00276310
InChI Key BYYFEADAQZCIGM-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepine-2-carboxylic acid, 2h-1,5-benzodioxepin-2-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepine-2-carboxylic acid, 2h-1,5-benzodioxepin-2-carboxylic acid, 3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylicacid
PubChem CID 53708627
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-4-carboxylic acid
SMILES C1COC2=CC=CC=C2OC1C(=O)O

Specifications

Melting Point 146°C to 148°C
Quantity 5 g
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only