missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine, 97%

Catalog No. p-7032246
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3364103 1 g
AAH3364106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3364103 Supplier Thermo Scientific Chemicals Supplier No. H3364103

CAS: 5735-53-5 | C8H9NO | 135.166 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5735-53-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD02181098
InChI Key YRLORWPBJZEGBX-UHFFFAOYSA-N
Synonym benzomorpholine, 3,4-dihydro-2h-benzo b 1,4 oxazine, 3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro, 2,3-dihydro-1,4-benzoxazine, akos bc-1149, 3,4-dihydro-2h-1,4-benzoxazin, pubchem22677, acmc-1amxt, 3,4-dihydro-2h-1,4-benzooxazine
PubChem CID 585096
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC=CC=C2N1

Specifications

Boiling Point 85°C (1 mmHg)
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzoxazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only