missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific Chemicals

Catalog No. p-7054242
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6229003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6229003 Supplier Thermo Scientific Chemicals Supplier No. H6229003

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1155264-46-2
Molecular Formula C14H20BNO3
Molecular Weight (g/mol) 261.13
MDL Number MFCD18073255
InChI Key HFUHUNYUUCDCAU-UHFFFAOYSA-N
Synonym 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, acmc-2099qf, 3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine
PubChem CID 54759084
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1

Specifications

Quantity 1 g
Formula Weight 261.12
Physical Form Solid
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only